CAS RN 38934-20-2
CAS RN 38934-20-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Quercetin 7-O-Beta-D-Glucuronide, TRC
CAS: 38934-20-2 Molecular Formula: C21H18O13 Molecular Weight (g/mol): 478.36 Synonym: 2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-4-oxo-4H-1-benzopyran-7-yl β-D-Glucopyranosiduronic Acid,Quercetin 7-O-β-Glucuronide,Quercetin 7-Glucuronide; SMILES: C1=CC(=C(C=C1C2=C(C(=O)C3=C(C=C(C=C3O2)O[C@H]4[C@@H]([C@H]([C@@H]([C@H](O4)C(=O)O)O)O)O)O)O)O)O