CAS RN 38934-69-9
CAS RN 38934-69-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Beta-D-Ribofuranosyl-1,2,4-triazole-3-carboxylic Acid Methyl Ester (Ribavirin Impurity H), TRC
CAS: 38934-69-9 Molecular Formula: C9 H13 N3 O7 Molecular Weight (g/mol): 275.22 Synonym: 1-β-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic Acid Methyl Ester,Methyl 1-(β-D-Ribofuranosyl)-1,2,4-triazole-3-carboxylate,NSC 360639 IUPAC Name: [(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl] 1H-1,2,4-triazol-3-ylmethyl carbonate SMILES: OC[C@H]1O[C@@H](OC(=O)OCc2nc[nH]n2)[C@H](O)[C@@H]1O