accessibility menu, dialog, popup

UserName

P

CAS RN 39319-11-4

IUPAC Name: triphenylphosphane
Synonyms: triphenylphosphine
Molecular Weight (g/mol): 262.29
Molecular Formula: C18H15P
InChi Key: RIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)