accessibility menu, dialog, popup

UserName

CH3H3COOH3COOCH3

CAS RN 39520-19-9

IUPAC Name: 1,3-dimethyl 2-(pentan-3-yl)propanedioate
Synonyms: 1,3-dimethyl 2-(pentan-3-yl)propanedioate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: LHJTVXUTWGDTQT-UHFFFAOYSA-N
SMILES: CCC(CC)C(C(=O)OC)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results