accessibility menu, dialog, popup

UserName

CH3H3CCH3OOCH3OOH3C

CAS RN 39520-25-7

IUPAC Name: 1,3-dimethyl 2-tert-butylpropanedioate
Synonyms: 1,3-dimethyl 2-tert-butylpropanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: OBANUQSDPDUGSL-UHFFFAOYSA-N
SMILES: COC(=O)C(C(=O)OC)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results