accessibility menu, dialog, popup

UserName

CH3NIN

CAS RN 39806-90-1

IUPAC Name: 4-iodo-1-methyl-1H-pyrazole
Synonyms: 4-iodo-1-methylpyrazole
Molecular Weight (g/mol): 208
Molecular Formula: C4H5IN2
InChi Key: RSDRDHPLXWMTRJ-UHFFFAOYSA-N
SMILES: CN1C=C(I)C=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)