accessibility menu, dialog, popup

UserName

CAS RN 39842-01-8

IUPAC Name: 1-isothiocyanato-2,4-dimethylbenzene
Synonyms: 1-isothiocyanato-2,4-dimethylbenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: HOHSBFCSOARUBF-UHFFFAOYSA-N
SMILES: CC1=CC=C(N=C=S)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results