accessibility menu, dialog, popup

UserName

H3CONO

CAS RN 39910-98-0

IUPAC Name: 1-[4-(morpholin-4-yl)phenyl]ethan-1-one
Synonyms: acetophenone, 4'-morpholino-
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: AKQWEDMTPCAESO-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)N1CCOCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results