accessibility menu, dialog, popup

UserName

HOONF

CAS RN 402-65-3

IUPAC Name: 2-fluoropyridine-4-carboxylic acid
Synonyms: 2-fluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: JMPFWDWYGOWUFP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results