accessibility menu, dialog, popup

UserName

BrOOO

CAS RN 40288-65-1

IUPAC Name: 1-(2H-1,3-benzodioxol-5-yl)-2-bromoethan-1-one
Synonyms: 1-(2H-1,3-benzodioxol-5-yl)-2-bromoethanone
Molecular Weight (g/mol): 243.06
Molecular Formula: C9H7BrO3
InChi Key: QBXCVQVFPVXAGS-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C2OCOC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results