accessibility menu, dialog, popup

UserName

CH3ClNCO

CAS RN 40397-90-8

IUPAC Name: 1-chloro-3-isocyanato-2-methylbenzene
Synonyms: 1-chloro-3-isocyanato-2-methylbenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: NGQMCUWZGVMILT-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1Cl)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results