accessibility menu, dialog, popup

UserName

CAS RN 40397-95-3

NOOClNCO

CAS RN 40397-95-3

IUPAC Name: 2-chloro-1-isocyanato-4-nitrobenzene
Synonyms: 2-chloro-1-isocyanato-4-nitrobenzene
Molecular Weight (g/mol): 198.56
Molecular Formula: C7H3ClN2O3
InChi Key: POLDSAUWRJCSOK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results