accessibility menu, dialog, popup

UserName

OCNN

CAS RN 40465-45-0

IUPAC Name: 4-isocyanatobenzonitrile
Synonyms: 4-isocyanatobenzonitrile
Molecular Weight (g/mol): 144.13
Molecular Formula: C8H4N2O
InChi Key: TVSXDZNUTPLDKY-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(C=C1)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results