accessibility menu, dialog, popup

UserName

CAS RN 405224-23-9

NBrClN

CAS RN 405224-23-9

IUPAC Name: 5-bromo-2-chloropyridine-3-carbonitrile
Synonyms: 5-bromo-2-chloropyridine-3-carbonitrile
Molecular Weight (g/mol): 217.45
Molecular Formula: C6H2BrClN2
InChi Key: MQOHJAYYYVQBSH-UHFFFAOYSA-N
SMILES: ClC1=NC=C(Br)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results