accessibility menu, dialog, popup

UserName

H3COOCH3NCS

CAS RN 40532-06-7

IUPAC Name: 2-isothiocyanato-1,4-dimethoxybenzene
Synonyms: 2-isothiocyanato-1,4-dimethoxybenzene
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: RADQLZMDTYWZKA-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)N=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)