accessibility menu, dialog, popup

UserName

CAS RN 40576-21-4

OHNN

CAS RN 40576-21-4

IUPAC Name: 3-(piperidine-1-carbonyl)piperidine
Synonyms: 3-(piperidine-1-carbonyl)piperidine
Molecular Weight (g/mol): 196.29
Molecular Formula: C11H20N2O
InChi Key: OSZRYTYCKGZYLB-UHFFFAOYNA-N
SMILES: O=C(C1CCCNC1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results