CAS RN 405939-39-1
IUPAC Name:
tert-butyl N-(5-bromo-1,3-thiazol-2-yl)carbamate
Synonyms:
tert-butyl N-(5-bromo-1,3-thiazol-2-yl)carbamate
Molecular Weight (g/mol):
279.15
Molecular Formula:
C8H11BrN2O2S
InChi Key:
OIBKBVFFZYCBAQ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=NC=C(Br)S1
1
–
1
of
1
results
1
2-(Boc-amino)-5-bromothiazole, 97%, Thermo Scientific™
CAS: 405939-39-1 Molecular Formula: C8H11BrN2O2S Molecular Weight (g/mol): 279.152 MDL Number: MFCD07368614 InChI Key: OIBKBVFFZYCBAQ-UHFFFAOYSA-N Synonym: n-boc-2-amino-5-bromothiazole,tert-butyl 5-bromothiazol-2-yl carbamate,tert-butyl 5-bromothiazol-2-ylcarbamate,tert-butyl n-5-bromo-1,3-thiazol-2-yl carbamate,5-bromo-thiazol-2-yl-carbamic acid tert-butyl ester,tert-butyl 5-bromo-1,3-thiazol-2-yl carbamate,carbamic acid, 5-bromo-2-thiazolyl-, 1,1-dimethylethyl ester,pubchem8907,n-boc-2-amno-5-bromothazole,tert-butyl-5-bromothiazol-2-ylcarbamate PubChem CID: 21300388 IUPAC Name: tert-butyl N-(5-bromo-1,3-thiazol-2-yl)carbamate SMILES: CC(C)(C)OC(=O)NC1=NC=C(S1)Br