accessibility menu, dialog, popup

UserName

H3COOI

CAS RN 40596-44-9

IUPAC Name: 4-iodobutyl acetate
Synonyms: 4-iodobutyl acetate
Molecular Weight (g/mol): 242.06
Molecular Formula: C6H11IO2
InChi Key: FLPVVTDEKLGZDZ-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results