CAS RN 406476-31-1
IUPAC Name:
2-(4-carboxylatopiperidin-1-yl)-5-(trifluoromethyl)pyridin-1-ium
Synonyms:
2-(4-carboxylatopiperidin-1-yl)-5-(trifluoromethyl)pyridin-1-ium
Molecular Weight (g/mol):
274.24
Molecular Formula:
C12H13F3N2O2
InChi Key:
KNDSIDUPVUCATQ-UHFFFAOYSA-N
SMILES:
[O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F
1
–
1
of
1
results
1
1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™
CAS: 406476-31-1 Molecular Formula: C12H13F3N2O2 Molecular Weight (g/mol): 274.24 MDL Number: MFCD00209248 InChI Key: KNDSIDUPVUCATQ-UHFFFAOYSA-N Synonym: 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylic acid,1-5-trifluoromethyl-2-pyridyl piperidine-4-carboxylic acid,5'-trifluoromethyl-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4-carboxylic acid,peakdale1_002677,2-4-carboxypiperidin-1-yl-5-trifluoromethyl pyridine,4-carboxy-1-5-trifluoromethyl pyridin-2-yl piperidine,1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylicacid,1-5-trifluoromethylpyrid-2-yl-piperidine-4-carboxylic acid PubChem CID: 2777788 SMILES: [O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F