CAS RN 40709-69-1
CAS RN 40709-69-1
IUPAC Name:
(5S)-6,6-dimethyl-5-[(10R)-12-oxo-3,5,11-trioxatricyclo[7.3.0.0²,⁶]dodeca-1,6,8-trien-10-yl]-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium iodide
Synonyms:
(5S)-6,6-dimethyl-5-[(10R)-12-oxo-3,5,11-trioxatricyclo[7.3.0.0²,⁶]dodeca-1,6,8-trien-10-yl]-2H,5H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium iodide
Molecular Weight (g/mol):
509.3
Molecular Formula:
C21H20INO6
InChi Key:
HKJKCPKPSSVUHY-GRTNUQQKSA-M
SMILES:
[I-].C[N+]1(C)CCC2=CC3=C(OCO3)C=C2[C@H]1[C@@H]1OC(=O)C2=C3OCOC3=CC=C12
1
–
2
of
2
results
2
Promotions Available
Tocris Bioscience™ (-)-Bicuculline methiodide
Tocris Bioscience™ (-)-Bicuculline methiodide
CAS: 40709-69-1 Molecular Formula: C21H20INO6