accessibility menu, dialog, popup

UserName

CH3OPONCOH3C

CAS RN 41003-94-5

IUPAC Name: diethyl (isocyanomethyl)phosphonate
Synonyms: diethyl isocyanomethylphosphonate
Molecular Weight (g/mol): 177.14
Molecular Formula: C6H12NO3P
InChi Key: ICURVXBGGDVHDT-UHFFFAOYSA-N
SMILES: CCOP(=O)(C[N+]#[C-])OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results