accessibility menu, dialog, popup

UserName

CAS RN 41019-45-8

IUPAC Name: 5-(4-chlorophenyl)furan-2-carboxylate
Synonyms: 5-(4-chlorophenyl)furan-2-carboxylate
Molecular Weight (g/mol): 221.62
Molecular Formula: C11H6ClO3
InChi Key: XIPQHWUSDHTXOO-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(O1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results