accessibility menu, dialog, popup

UserName

CAS RN 4105-92-4

CH3OOOOCH3OOCH3

CAS RN 4105-92-4

IUPAC Name: 1,3,5-triethyl benzene-1,3,5-tricarboxylate
Synonyms: 1,3,5-triethyl benzene-1,3,5-tricarboxylate
Molecular Weight (g/mol): 294.3
Molecular Formula: C15H18O6
InChi Key: KXGOWZRHSOJOLF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results