CAS RN 41223-27-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-methoxybutan-2-ol, TRC
CAS: 41223-27-2 Molecular Formula: C5H12O2 Molecular Weight (g/mol): 104.15 IUPAC Name: 4-methoxybutan-2-ol SMILES: COCCC(C)O