CAS RN 4138-26-5
IUPAC Name:
(3R)-3-carbamoylpiperidin-1-ium
Synonyms:
(R)-nipecotamide(1+) (3R)-piperidine-3-carboxamide (R)-nipecotamide cation (3R)-hexahydronicotinamide(1+)
Molecular Weight (g/mol):
129.18
Molecular Formula:
C6H13N2O
InChi Key:
BVOCPVIXARZNQN-RXMQYKEDSA-O
SMILES:
NC(=O)[C@@H]1CCC[NH2+]C1
1
–
1
of
1
results
1
Nipecotamide, 95%, Thermo Scientific Chemicals
CAS: 4138-26-5 Molecular Formula: C6H12N2O Molecular Weight (g/mol): 128.17 MDL Number: MFCD00005993 InChI Key: BVOCPVIXARZNQN-UHFFFAOYSA-N Synonym: nipecotamide,3-piperidinecarboxamide,nipecotic acid amide,hexahydronicotinamide,piperidine-3-carboxylic acid amide,racemic nipecotamide,pubchem11797,acmc-20lak2,3-piperidine carboxamide PubChem CID: 92980 ChEBI: CHEBI:60116 IUPAC Name: piperidine-3-carboxamide SMILES: C1CC(CNC1)C(=O)N