accessibility menu, dialog, popup

UserName

ONOH

CAS RN 41536-44-1

IUPAC Name: 2-(morpholin-4-yl)phenol
Synonyms: 2-(morpholin-4-yl)phenol
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: VMPYFWTYGZZUMY-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results