accessibility menu, dialog, popup

UserName

HPBH3

CAS RN 41593-58-2

IUPAC Name: diphenylphosphane boron
Synonyms: diphenylphosphine boron
Molecular Weight (g/mol): 197
Molecular Formula: C12H11BP
InChi Key: LRNNYBXLESRLOS-UHFFFAOYSA-N
SMILES: [B].P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)