CAS RN 41906-86-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nitrocefin (>90%), TRC
CAS: 41906-86-9 Molecular Formula: C21 H16 N4 O8 S2 Molecular Weight (g/mol): 516.5 Synonym: (6R,7R)-3-[(1E)-2-(2,4-dinitrophenyl)ethenyl]-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid,[6R-[3(E),6α,7β]]-3-[2-(2,4-dinitrophenyl)ethenyl]-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid; IUPAC Name: (6R,7R)-7-[[(E)-4-(2,4-dinitrophenyl)-2-thiophen-2-ylbut-3-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid SMILES: OC(=O)C1=CCS[C@@H]2[C@H](NC(=O)C(\C=C\c3ccc(cc3[N+](=O)[O-])[N+](=O)[O-])c4cccs4)C(=O)N12