accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 4208-64-4

IUPAC Name: 1-(furan-2-yl)ethan-1-ol
Synonyms: 2-furanmethanol, α-methyl-
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: UABXUIWIFUZYQK-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CO1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)