accessibility menu, dialog, popup

UserName

H3CMgCl

CAS RN 42092-01-3

IUPAC Name: magnesium(2+) heptan-1-ide chloride
Synonyms: magnesium(2+) heptan-1-ide chloride
Molecular Weight (g/mol): 158.95
Molecular Formula: C7H15ClMg
InChi Key: YBORTXLIZPFKPR-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CCCCCC[CH2-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results