CAS RN 42105-98-6
CAS RN 42105-98-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(methylamino)-3-oxopropanoic acid, TRC
CAS: 42105-98-6 Molecular Formula: C4H7NO3 Molecular Weight (g/mol): 117.1 IUPAC Name: 3-(methylamino)-3-oxopropanoic acid SMILES: CNC(=O)CC(=O)O