accessibility menu, dialog, popup

UserName

CAS RN 42327-95-7

H3COOHOH3C

CAS RN 42327-95-7

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid
Synonyms: 2,2-dimethyl-3H-1-benzofuran-7-carboxylic acid
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: AVVACPBRCFXZMR-UHFFFAOYSA-N
SMILES: CC1(C)CC2=CC=CC(C(O)=O)=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results