accessibility menu, dialog, popup

UserName

CAS RN 42330-10-9

H3COCH3Br

CAS RN 42330-10-9

IUPAC Name: 3-bromoheptan-4-one
Synonyms: 3-bromoheptan-4-one
Molecular Weight (g/mol): 193.08
Molecular Formula: C7H13BrO
InChi Key: SFKVBRLKXVRUQW-UHFFFAOYNA-N
SMILES: CCCC(=O)C(Br)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results