accessibility menu, dialog, popup

UserName

OHOSHN

CAS RN 42607-21-6

IUPAC Name: 2-phenyl-1,3-thiazolidine-4-carboxylic acid
Synonyms: 2-phenyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 209.26
Molecular Formula: C10H11NO2S
InChi Key: AZDYQBFYMBALBY-UHFFFAOYNA-N
SMILES: OC(=O)C1CSC(N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results