CAS RN 4282-78-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Propargyl Phenothiazine, TRC
CAS: 4282-78-4 Molecular Formula: C15H11NS Molecular Weight (g/mol): 237.32 Synonym: 10-Propargylphenothiazine,10-(2-Propyn-1-yl)-10H-phenothiazine IUPAC Name: 10-prop-2-ynylphenothiazine SMILES: C#CCN1c2ccccc2Sc3ccccc13