accessibility menu, dialog, popup

UserName

CAS RN 42885-14-3

CH3NN

CAS RN 42885-14-3

IUPAC Name: 5-methylpyridine-3-carbonitrile
Synonyms: 5-methylpyridine-3-carbonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: VDWQWOXVBXURMT-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=C1)C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results