CAS RN 42923-77-3
IUPAC Name:
6-methoxy-1,2,3,4-tetrahydroisoquinoline
Synonyms:
6-methoxy-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol):
163.22
Molecular Formula:
C10H13NO
InChi Key:
YYTAYINRPUJPNH-UHFFFAOYSA-N
SMILES:
COC1=CC=C2CNCCC2=C1
1
–
1
of
1
results
1
6-Methoxy-1,2,3,4-tetrahydroisoquinoline, TRC
CAS: 42923-77-3 Molecular Formula: C10H13NO Molecular Weight (g/mol): 163.22 Synonym: 1,2,3,4-Tetrahydro-6-methoxyisoquinoline,6-Methoxy-1,2,3,4-tetrahydroisoquinoline IUPAC Name: 6-methoxy-1,2,3,4-tetrahydroisoquinoline SMILES: COC1=CC=C(CNCC2)C2=C1