accessibility menu, dialog, popup

UserName

CAS RN 42959-38-6

IUPAC Name: 2-chloro-5-nitropyridine-3-carboxylic acid
Synonyms: 2-chloro-5-nitropyridine-3-carboxylic acid
Molecular Weight (g/mol): 202.55
Molecular Formula: C6H3ClN2O4
InChi Key: WOFZRBCITDVRON-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CN=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results