accessibility menu, dialog, popup

UserName

CAS RN 4301-14-8

HCMgBr

CAS RN 4301-14-8

IUPAC Name: magnesium(2+) acetylene bromide
Synonyms: magnesium(2+) acetylene bromide
Molecular Weight (g/mol): 129.24
Molecular Formula: C2HBrMg
InChi Key: LROBJRRFCPYLIT-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C#[C-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results