accessibility menu, dialog, popup

UserName

CAS RN 431942-30-2

NOBr

CAS RN 431942-30-2

IUPAC Name: 5-bromo-2-phenylfuro[2,3-b]pyridine
Synonyms: 5-bromo-2-phenylfuro[2,3-b]pyridine
Molecular Weight (g/mol): 274.12
Molecular Formula: C13H8BrNO
InChi Key: GDEUCQHGCNNNOT-UHFFFAOYSA-N
SMILES: BrC1=CN=C2OC(=CC2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results