accessibility menu, dialog, popup

UserName

H3CCH3CH3Cl

CAS RN 4325-50-2

IUPAC Name: 2-chloro-2-methylnonane
Synonyms: 2-chloro-2-methylnonane
Molecular Weight (g/mol): 176.73
Molecular Formula: C10H21Cl
InChi Key: QDJXQCKHBZEIJM-UHFFFAOYSA-N
SMILES: CCCCCCCC(C)(C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)