CAS RN 4328-94-3
CAS RN 4328-94-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3,5-Pentanetriol, TRC
CAS: 4328-94-3 Molecular Formula: C5H12O3 Molecular Weight (g/mol): 120.15 IUPAC Name: pentane-1,3,5-triol SMILES: OCCC(O)CCO