CAS RN 4329-75-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Diisonicotinoylhydrazine, TRC
CAS: 4329-75-3 Molecular Formula: C12 H10 N4 O2 Molecular Weight (g/mol): 242.23 Synonym: Isonicotinic Acid N'-(Pyridine-4-carbonyl)hydrazide,N,N'-Bis(4-picolinoyl)hydrazine,N,N'-Diisonicotinoylhydrazine,NSC 39301,NSC 52961 IUPAC Name: N'-(pyridine-4-carbonyl)pyridine-4-carbohydrazide SMILES: O=C(NNC(=O)c1ccncc1)c2ccncc2