CAS RN 4342-03-4
CAS RN 4342-03-4
IUPAC Name:
(4E)-4-(3,3-dimethyltriazan-1-ylidene)-4H-imidazole-5-carboxamide
Synonyms:
dacarbazine
Molecular Weight (g/mol):
182.19
Molecular Formula:
C6H10N6O
InChi Key:
OMJKFYKNWZZKTK-UXBLZVDNSA-N
SMILES:
CN(C)N\N=C1\N=CN=C1C(N)=O
1
–
1
of
1
results
1
Dacarbazine, Thermo Scientific™
CAS: 4342-03-4 Molecular Formula: C4H3IN2O Molecular Weight (g/mol): 221.99 MDL Number: MFCD00057167 InChI Key: SMIGOGPUTQXOKF-UHFFFAOYSA-N Synonym: 5-iodopyrimidin-4-ol,5-iodopyrimidin-4 3h-one,4-hydroxy-5-iodopyrimidine,5-iodo-1h-pyrimidin-4-one,4 1h-pyrimidinone, 5-iodo,5-iodopyrimidin-4 1h-one,5-iodo-3h-pyrimidin-4-one,5-iodo-3,4-dihydropyrimidin-4-one,5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one PubChem CID: 12356582 IUPAC Name: 5-iodo-1H-pyrimidin-6-one SMILES: C1=C(C(=O)NC=N1)I