accessibility menu, dialog, popup

UserName

CAS RN 4369-14-6

H3COSiOOCH2OCH3OCH3

CAS RN 4369-14-6

IUPAC Name: 3-(trimethoxysilyl)propyl prop-2-enoate
Synonyms: acryloyloxy propyltrimethoxylane acryloyloxy propyltrimethoxysilane
Molecular Weight (g/mol): 234.32
Molecular Formula: C9H18O5Si
InChi Key: KBQVDAIIQCXKPI-UHFFFAOYSA-N
SMILES: CO[Si](CCCOC(=O)C=C)(OC)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results