CAS RN 4376-19-6
CAS RN 4376-19-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Monopropyl Phthalate, TRC
CAS: 4376-19-6 Molecular Formula: C11H12O4 Molecular Weight (g/mol): 208.21 Synonym: 1,2-Benzenedicarboxylic Acid 1-Propyl Ester,1,2-Benzenedicarboxylic Acid Monopropyl Ester,Phthalic Acid Monopropyl Ester,Phthalic Acid Propyl Ester,Mono-n-propyl phthalate,NSC 309966 IUPAC Name: 2-propoxycarbonylbenzoic acid SMILES: CCCOC(=O)C1=CC=CC=C1C(=O)O