accessibility menu, dialog, popup

UserName

CH3NOClFONH2(?)

CAS RN 4415-11-6

IUPAC Name: 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxamide
Synonyms: 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxamide
Molecular Weight (g/mol): 254.65
Molecular Formula: C11H8ClFN2O2
InChi Key: PPKCMGUTHICRLU-UHFFFAOYSA-N
SMILES: CC1=C(C(N)=O)C(=NO1)C1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results