accessibility menu, dialog, popup

UserName

HCNN

CAS RN 442517-33-1

IUPAC Name: 6-ethynylquinoxaline
Synonyms: 6-ethynylquinoxaline
Molecular Weight (g/mol): 154.17
Molecular Formula: C10H6N2
InChi Key: QSYDWUWZZBZOCD-UHFFFAOYSA-N
SMILES: C#CC1=CC=C2N=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)