CAS RN 443-82-3
IUPAC Name:
1-fluoro-2,3-dimethylbenzene
Synonyms:
1-fluoro-2,3-dimethylbenzene
Molecular Weight (g/mol):
124.16
Molecular Formula:
C8H9F
InChi Key:
AWLDSXJCQWTJPC-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(F)=C1C
1
–
1
of
1
results
1
3-Fluoro-o-xylene, 99%
CAS: 443-82-3 Molecular Formula: C8H9F Molecular Weight (g/mol): 124.16 MDL Number: MFCD00000323 InChI Key: AWLDSXJCQWTJPC-UHFFFAOYSA-N Synonym: 3-fluoro-o-xylene,2,3-dimethylfluorobenzene,3-fluoro-1,2-xylene,o-xylene, 3-fluoro,3-fluoro-1,2-dimethylbenzene,benzene, 1-fluoro-2,3-dimethyl,1,2-dimethyl-3-fluorobenzene,1-fluoro-2,3-dimethyl-benzene,fluorxylol,pubchem4408 PubChem CID: 96489 IUPAC Name: 1-fluoro-2,3-dimethylbenzene SMILES: CC1=CC=CC(F)=C1C