accessibility menu, dialog, popup

UserName

NH2

CAS RN 4442-85-7

IUPAC Name: 2-cyclohexylethan-1-amine
Synonyms: cyclohexaneethanamine (9CI)
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: HFACYWDPMNWMIW-UHFFFAOYSA-N
SMILES: NCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results